Distilled Water
Gibco™

Distilled Water

This distilled water is high-quality water for use as a solvent in the preparation of cell culture media and laboratoryRead more
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
10 L
20 L
Catalog number 15230261
Price (USD)
313.00
Each
Quantity:
10 L
Customize this product
Price (USD)
313.00
Each
This distilled water is high-quality water for use as a solvent in the preparation of cell culture media and laboratory reagents. Available in many sizes to meet your needs, this distilled water features:

• Cell culture quality
• Proven performance
• Suitable for use in cell culture and molecular biology applications

Cell culture quality
This water is prepared by distillation and filtration (0.1 μm) with no added substances. Multiple aspects of water quality are monitored on a routine basis: conductivity, total organic carbon (TOC), bioburden, endotoxin, and metals.

Proven performance
The performance and consistency expected from Gibco™ liquid media products depends on the high-quality water we use in manufacturing. This water is also bottled as Gibco™ Distilled Water.

Use in cell culture and molecular biology applications
This distilled water can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions, and diluting buffers.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
PackagingUniversal Bag
Product LineGibco
Purification MethodMembrane-Filtered
Recommended StorageStorage conditions: 15 to 30°C
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Shipping ConditionRoom Temperature
SterilitySterile-filtered
Treatment(s)Not DEPC-Treated
GradeCell Culture
Quantity10 L
Unit SizeEach

Frequently asked questions (FAQs)

Can I get the conductivity measurement for Distilled Water?

We do not test the “finished” product for conductivity as part of the QC testing specifications. However, conductivity testing is done in our water system on a daily basis. This is the water system used in the manufacture of cell culture media and reagents made at the Gibco site.

Conductivity testing is performed according to USP specifications. The USP specification is that conductivity be “not more than 1 micromho/cm”.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the difference between Water for Injection (WFI) for Cell Culture, Cat. No. A12873XX and Distilled Water, Cat. No. 15230XXX?

These two products are the same water. However, Water for Injection (WFI) for Cell Culture has undergone more testing, including conductivity measurement as well as testing for nitrate and total organic carbon.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Will your company continue to support other films?

Yes. Although we feel that the PE film exceeds many of the technical properties of the other available films, we will continue to offer and support our other currently available films through custom orders.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

In what size package will my Gibco Cell Culture Bags ship?

Each bag size will arrive in a specific heavyweight corrugated box designed for shipment:
5 L: 12” x 14.25” x 5.5”
10 L: 13” x 23.5” x 6.125”
20 L: 15” x 24” x 6.125”

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What are the benefits of the Gibco PE film?

The commercially available film is a very clear, multi-layered, single web film with a polyethylene contact surface that can be made into a wide range of bag sizes (250 mL to 1,000L). In addition, the film is a market leader with exceptional gas barrier properties and very low extractable and leachable results. Click here to read more about the PE Film specifications (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/classical-media/gibco-cell-culture-bags.html).

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.