1,2-Diaminopropane, 99%
1,2-Diaminopropane, 99%
1,2-Diaminopropane, 99%
Thermo Scientific Chemicals

1,2-Diaminopropane, 99%

CAS: 78-90-0 | C3H10N2 | 74.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1524000101 LGlass bottle
Catalog number 152400010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS78-90-0
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Clear liquid
Refractive index1.4450 to 1.4470 (20°C, 589 nm)
Color scale=<20 APHA
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only