1,2-Diaminopropane, 99%
1,2-Diaminopropane, 99%
1,2-Diaminopropane, 99%
Thermo Scientific Chemicals

1,2-Diaminopropane, 99%

CAS: 78-90-0 | C3H10N2 | 74.13 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
產品號碼 152400025
價格 (TWD)
-
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS78-90-0
規格Specification Sheet規格表
Infrared spectrumConforms
Appearance (Form)Clear liquid
Refractive index1.4450 to 1.4470 (20°C, 589 nm)
Color scale=<20 APHA
GC>=98.5 %
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only