Antibiotic-Antimycotic (100X)
Gibco™

Antibiotic-Antimycotic (100X)

Solution contains penicillin, streptomycin, and Gibco™ Amphotericin B to prevent cell culture contamination.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
1524009620 mL
15240062100 mL
1524011220 x 100 mL
Catalog number 15240096
Price (BRL)
146,74
Each
Quantity:
20 mL
Price (BRL)
146,74
Each

Gibco™ Antibiotic-Antimycotic is used to prevent bacterial and fungal contamination. This solution contains 10,000 units/mL of penicillin, 10,000 μg/mL of streptomycin, and 25 μg/mL of Gibco™ Amphotericin B. The antibiotics penicillin and streptomycin prevent bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria. Amphotericin B prevents fungal contamination of cell cultures due to its inhibition of multi-cellular fungus and yeast.

Penicillin was originally purified from the fungus Penicillium and acts by interfering directly with the turnover of the bacteria cell wall and indirectly by triggering the release of enzymes that further alter the cell wall. Streptomycin was originally purified from Streptomyces griseus. It acts by binding to the 30S subunit of the bacterial ribosome leading to inhibition of protein synthesis and death in susceptible bacteria. Amphotericin B is an antifungal agent that prevents the growth of fungi and yeast by causing an increase in fungal plasma membrane permeability.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Concentration100 X
Culture TypeMammalian Cell Culture
Quantity20 mL
Shelf Life12 Months
Shipping ConditionDry Ice
FormLiquid
Product TypeAntibiotic-Antimycotic
SterilitySterile-filtered
Sterilization MethodSterile-filtered
Unit SizeEach
Contents & Storage
Storage conditions: -5 to -20°C
Shipping conditions: Dry ice
Shelf life: 12 months from date of manufacture

Frequently asked questions (FAQs)

Can I add antibiotics to OptiPRO SFM?

You can supplement OptiPRO SFM with 5mL/L Antibiotic-Antimycotic (Cat. No. 15240096)

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Can I use antibiotics such as Pen-Strep or Antibiotic-Antimycotic (100X) when culturing Sf9 insect cells?

Yes, however, we do not recommend adding antibiotics to the medium at the time of thawing. Thaw cells into medium without antibiotic and allow the cells to recover from the thawing process. After that, you can add antibiotics by diluting into the medium.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the solvent used in Antibiotic-Antimycotic (100X)?

The components are in a 0.85% saline solution.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What are the components of Antibiotic-Antimycotic (100X)?

This solution contains 10,000 units/mL of penicillin, 10,000 µg/mL of streptomycin, and 25 µg/mL of Gibco Amphotericin B.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the recommended storage condition for Antibiotic-Antimycotic (100X)?

The recommended condition is storage at -5 to -20 degrees C and when stored as recommended, the shelf life is 12 months from the date of shipment.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (6)

Citations & References
Abstract
Bombyxin is a growth factor for wing imaginal disks in Lepidoptera.
Authors: Nijhout H Frederik; Grunert Laura W;
Journal:Proc Natl Acad Sci U S A
PubMed ID:12429853
'The mechanisms that control the growth rate of internal tissues during postembryonic development are poorly understood. In insects, the growth rate of imaginal disks varies with nutrition and keeps pace with variation in somatic growth. We describe here a mechanism by which the growth of wing imaginal disks is controlled. ... More
Apoptosis induces efflux of the mitochondrial matrix enzyme deoxyguanosine kinase.
Authors:Jüllig M, Eriksson S,
Journal:J Biol Chem
PubMed ID:11294860
Deoxyguanosine kinase (dGK) initiates the salvage of purine deoxynucleosides in mitochondria and is a key enzyme in mitochondrial DNA precursor synthesis. The active form of the enzyme is a 60-kDa protein normally located in the mitochondrial matrix. Here we describe the subcellular distribution of dGK during apoptosis in human epithelial ... More
Hypoxia and Nitric Oxide Treatment Confer Tolerance to Glucose Starvation in a 5'-AMP-activated Protein Kinase-dependent Manner.
Authors: Esumi Hiroyasu; Izuishi Kunihiko; Kato Kazuyoshi; Hashimoto Koichi; Kurashima Yukiko; Kishimoto Atsuhiro; Ogura Tsutomu; Ozawa Takayuki;
Journal:J Biol Chem
PubMed ID:12091379
Hypoxia is a critical event for higher organisms, and cells and tissues react by increasing the oxygen supply by vasodilatation, angiogenesis, and erythropoiesis and maintaining cellular energy by increasing glycolysis and inhibiting anabolic pathways. Stimulation of glycolysis has been regarded as the main response that increases energy production during hypoxia; ... More
Metabolic stress activates an ERK/hnRNPK/DDX3X pathway in pancreatic ß cells.
Authors:Good AL, Haemmerle MW, Oguh AU, Doliba NM, Stoffers DA
Journal:Mol Metab
PubMed ID:31178390
Pancreatic ß cell failure plays a central role in the development of type 2 diabetes (T2D). While the transcription factors shaping the ß cell gene expression program have received much attention, the post-transcriptional controls that are activated in ß cells during stress are largely unknown. We recently identified JUND as ... More
PNA Telomere and Centromere FISH Staining for Accurate Analysis of Radiation-Induced Chromosomal Aberrations.
Authors:Cartwright IM, Haskins JS, Kato TA
Journal:Methods Mol Biol
PubMed ID:31267424
Dicentric and centric ring chromosomes are used for radiation-induced damage analysis and biodosimetry after radiation exposure. However, Giemsa stain-based cytogenetic analysis is labor-intense and time-consuming. Moreover, the disadvantage of Giemsa based chromosome analysis is a potential poor reproducibility when researchers are not fully trained for analysis. These problems come from ... More