3-Aminocrotononitrile, 96%, mixture of cis and trans
3-Aminocrotononitrile, 96%, mixture of cis and trans
3-Aminocrotononitrile, 96%, mixture of cis and trans
3-Aminocrotononitrile, 96%, mixture of cis and trans
Thermo Scientific Chemicals

3-Aminocrotononitrile, 96%, mixture of cis and trans

CAS: 1118-61-2 | C4H6N2 | 82.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 152455000
Price (USD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1118-61-2
IUPAC Name(2E)-3-aminobut-2-enenitrile
Molecular FormulaC4H6N2
InChI KeyDELJOESCKJGFML-DUXPYHPUSA-N
SMILESC\C(N)=C/C#N
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=95 %
Appearance (Color)Yellow
Infrared spectrumConforms
Appearance (Form)Flakes
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only