trans-Anethole, 99%
trans-Anethole, 99%
trans-Anethole, 99%
Thermo Scientific Chemicals

trans-Anethole, 99%

CAS: 4180-23-8 | C10H12O | 148.2 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
產品號碼 152531000
價格 (TWD)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS4180-23-8
IUPAC Name1-methoxy-4-[(1E)-prop-1-en-1-yl]benzene
Molecular FormulaC10H12O
InChI KeyRUVINXPYWBROJD-ONEGZZNKSA-N
SMILESCOC1=CC=C(\C=C\C)C=C1
檢視更多
規格Specification Sheet規格表
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.5580 to 1.5650 (20°C, 589 nm)
GC>=98.5 %
Density(25/4°C) 0.980 to 0.985
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only