Rhodanine, 99%
Rhodanine, 99%
Rhodanine, 99%
Thermo Scientific Chemicals

Rhodanine, 99%

CAS: 141-84-4 | C3H3NOS2 | 133.183 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
產品號碼 152600250
價格 (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS141-84-4
IUPAC Name2-sulfanylidene-1,3-thiazolidin-4-one
Molecular FormulaC3H3NOS2
InChI KeyKIWUVOGUEXMXSV-UHFFFAOYSA-N
SMILESO=C1CSC(=S)N1
檢視更多
規格Specification Sheet規格表
Infrared spectrumConforms
Melting point167°C to 170°C
Titration with NaOH>=98.5 %
Appearance (Form)Crystalline powder
Appearance (Color)Light yellow to beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only