4-Fluorobenzoyl chloride, 98%
4-Fluorobenzoyl chloride, 98%
4-Fluorobenzoyl chloride, 98%
Thermo Scientific Chemicals

4-Fluorobenzoyl chloride, 98%

CAS: 403-43-0 | C7H4ClFO | 158.56 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
產品號碼 152951000
價格 (TWD)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS403-43-0
IUPAC Name4-fluorobenzoyl chloride
Molecular FormulaC7H4ClFO
InChI KeyCZKLEJHVLCMVQR-UHFFFAOYSA-N
SMILESFC1=CC=C(C=C1)C(Cl)=O
檢視更多
規格Specification Sheet規格表
GC>=97.5 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.5299 to 1.5319 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only