o-Vanillin, 99%
o-Vanillin, 99%
o-Vanillin, 99%
Thermo Scientific Chemicals

o-Vanillin, 99%

CAS: 148-53-8 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 153071000
Price (USD)
100.65
Special offer
Online exclusive
Ends: 30-Jun-2026
118.00
Save 17.35 (15%)
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
100.65
Special offer
Online exclusive
Ends: 30-Jun-2026
118.00
Save 17.35 (15%)
Each
Chemical Identifiers
CAS148-53-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals or flakes or crystalline mass
Infrared spectrumConforms
Melting point42°C to 45°C
Appearance (Color)Yellow to brown
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only