Neocuproine hemihydrate, 99+%
Neocuproine hemihydrate, 99+%
Neocuproine hemihydrate, 99+%
Thermo Scientific Chemicals

Neocuproine hemihydrate, 99+%

CAS: 34302-69-7 | C28H26N4O | 434.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 153320250
Price (EUR)
253,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
253,00
Each
Chemical Identifiers
CAS34302-69-7
IUPAC Namebis(2,9-dimethyl-1,10-phenanthroline) hydrate
Molecular FormulaC28H26N4O
InChI KeyIEBXFSLFDFHSRD-UHFFFAOYSA-N
SMILESO.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1.CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Melting point159°C to 164°C
Titration with HClO499.0 to 101.0 %
Appearance (Color)White to beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only