Aminoacetaldehyde dimethyl acetal, 99%
Aminoacetaldehyde dimethyl acetal, 99%
Aminoacetaldehyde dimethyl acetal, 99%
Thermo Scientific Chemicals

Aminoacetaldehyde dimethyl acetal, 99%

CAS: 22483-09-6 | C4H11NO2 | 105.14 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
產品號碼 153340025
價格 (TWD)
-
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS22483-09-6
IUPAC Name2,2-dimethoxyethan-1-amine
Molecular FormulaC4H11NO2
InChI KeyQKWWDTYDYOFRJL-UHFFFAOYSA-N
SMILESCOC(CN)OC
檢視更多
規格Specification Sheet規格表
GC>=98.5 %
Refractive index1.4160 to 1.4180 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only