m-Toluoyl chloride, 99%
m-Toluoyl chloride, 99%
m-Toluoyl chloride, 99%
Thermo Scientific Chemicals

m-Toluoyl chloride, 99%

CAS: 1711-06-4 | C8H7ClO | 154.6 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
產品號碼 153411000
價格 (TWD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS1711-06-4
IUPAC Name3-methylbenzoyl chloride
Molecular FormulaC8H7ClO
InChI KeyYHOYYHYBFSYOSQ-UHFFFAOYSA-N
SMILESCC1=CC=CC(=C1)C(Cl)=O
檢視更多
規格Specification Sheet規格表
Appearance (Form)Liquid
Infrared spectrumConforms
Appearance (Color)Clear colorless to light brown
Refractive index1.5475 to 1.5495 (20°C, 589 nm)
Titration RCOCl>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only