Appearance (Color)White to almost white to ochre to brown
Appearance (Form)Powder or crystalline powder and/or chunks
Infrared spectrumConforms
Melting point48°C to 53°C
Titration with HClO497.5 to 102.5 % (On dry substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.