Ferrocenecarboxaldehyde, 98%, Thermo Scientific Chemicals
Ferrocenecarboxaldehyde, 98%, Thermo Scientific Chemicals
Ferrocenecarboxaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ferrocenecarboxaldehyde, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 153530050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS12093-10-6
Molecular FormulaC11H10FeO
SMILES[Fe].c1cccc1.O=Cc1cccc1
Molecular Weight (g/mol)214.05
MDL NumberMFCD00001429
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to red to brown
GC>=97.5 %
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acylation reagent
RUO – Research Use Only