Plasmid pBR322
Invitrogen™

Plasmid pBR322

Plasmid pBR322 (1) contains three unique restriction endonuclease recognition sites within the !-lactamase (AmpR) gene and eight unique sites withinRead more
Have Questions?
Catalog NumberQuantity
1536701425 μg
Catalog number 15367014
Price (USD)
406.00
Each
Quantity:
25 μg
Price (USD)
406.00
Each
Plasmid pBR322 (1) contains three unique restriction endonuclease recognition sites within the !-lactamase (AmpR) gene and eight unique sites within the TetR gene. The plasmid, purified from DH10B™ E. coli, also has 14 non-selectable unique restriction endonuclease recognition sites. Unique cleavage sites for Hind III and Cla I are found in the promoter of the tetracycline resistance gene. Insertion of DNA at either of these two sites usually results in loss of tetracycline resistance.

Performance and Quality Testing: Purity, DNA structure, and selected restriction endonuclease fragmentation patterns are verified by agarose gel analysis. Transformation of E. coli HB101 to confirm AmpR and TetR is confirmed by plating on selective medium.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cloning MethodRestriction Enzyme/ MCS
Concentration0.25 μg/μl
For Use With (Application)Cloning
Product TypePlasmid
Quantity25 μg
Shipping ConditionApproved for shipment on Wet or Dry Ice
VectorpBR322
Unit SizeEach
Contents & Storage
pBR322 is supplied at 0.25 μg/μl in 10 mM Tris-HCl (pH 7.4), 5 mM NaCl, 0.1 mM EDTA. Store at +4°C.