2-Amino-5-bromopyridine, 97%
2-Amino-5-bromopyridine, 97%
2-Amino-5-bromopyridine, 97%
Thermo Scientific Chemicals

2-Amino-5-bromopyridine, 97%

CAS: 1072-97-5 | C5H5BrN2 | 173.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 153740050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1072-97-5
IUPAC Name5-bromopyridin-2-amine
Molecular FormulaC5H5BrN2
InChI KeyWGOLHUGPTDEKCF-UHFFFAOYSA-N
SMILESNC1=CC=C(Br)C=N1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Melting point135°C to 140°C
Appearance (Color)White to beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only