4-Aminohippuric acid, 99%
4-Aminohippuric acid, 99%
4-Aminohippuric acid, 99%
Thermo Scientific Chemicals

4-Aminohippuric acid, 99%

CAS: 61-78-9 | C9H10N2O3 | 194.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 153800250
Price (EUR)
107,00
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
107,00
Each
Chemical Identifiers
CAS61-78-9
IUPAC Name2-[(4-aminophenyl)formamido]acetic acid
Molecular FormulaC9H10N2O3
InChI KeyHSMNQINEKMPTIC-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1)C(=O)NCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white to light yellow
Appearance (Color)to light red to light brown
Appearance (Form)Powder
Infrared spectrumConforms
Melting point197°C to 204°C (Dec)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only