2-(4-Aminophenyl)ethylamine, 95%
2-(4-Aminophenyl)ethylamine, 95%
2-(4-Aminophenyl)ethylamine, 95%
Thermo Scientific Chemicals

2-(4-Aminophenyl)ethylamine, 95%

CAS: 13472-00-9 | C8H12N2 | 136.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
Packaging:
Glass bottle
Catalog number 153840050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13472-00-9
IUPAC Name4-(2-aminoethyl)aniline
Molecular FormulaC8H12N2
InChI KeyLNPMZQXEPNWCMG-UHFFFAOYSA-N
SMILESNCCC1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorlesss to yellow-brown
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5890 to 1.5930 (20°C, 589 nm)
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only