3-Aminophthalhydrazide, 98%, pure
3-Aminophthalhydrazide, 98%, pure
3-Aminophthalhydrazide, 98%, pure
Thermo Scientific Chemicals

3-Aminophthalhydrazide, 98%, pure

3-Aminophthalhydrazide, 98%, pure, Quantity: 25g, Packaging: Glass bottle, Blue-Purple to Green, Grade: Pure, Melting Point: 319.0 deg.C to 320.0 deg.C, Molecular Weight: 177.16, Percent Purity: 98%, Assay Percent Range: 98%, Beilstein: 25,IV,4192, CAS: 521-31-3, CAS Max %: 100.0 | CAS: 521-31-3 | C8H7N3O2 | 177.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 153851000
Price (TWD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS521-31-3
IUPAC Name5-amino-1,2,3,4-tetrahydrophthalazine-1,4-dione
Molecular FormulaC8H7N3O2
InChI KeyHWYHZTIRURJOHG-UHFFFAOYSA-N
SMILESNC1=C2C(=O)NNC(=O)C2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration excess NaOH>=97.5 %
Solubility(2 % in 1 N NaOH) Clear yellow to yellow-green
Appearance (Color)Yellow to yellow-green to beige
Appearance (Form)Powder and/or chunks
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only