2-Aminothiazole, 97%
2-Aminothiazole, 97%
2-Aminothiazole, 97%
Acros Organics

2-Aminothiazole, 97%

CAS: 96-50-4 | C3H4N2S | 100.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number 153895000
also known as 15389
HSN 29341000
Price (INR)
22,580.00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
22,580.00
Each
Chemical Identifiers
CAS96-50-4
IUPAC Name1,3-thiazol-2-amine
Molecular FormulaC3H4N2S
InChI KeyRAIPHJJURHTUIC-UHFFFAOYSA-N
SMILESNC1=NC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Brown
Appearance (Form)Powder or flakes or granules and/or lumps
Melting point86°C to 91°C
Titration with HClO4>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only