Anisole, 99%, pure
Anisole, 99%, pure
Anisole, 99%, pure
Thermo Scientific Chemicals

Anisole, 99%, pure

CAS: 100-66-3 | C7H8O | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 153920050
Price (USD)
48.20
Each
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
48.20
Each
Specifications
Assay Percent Range98.5% min. (GC)
Beilstein06,138
CAS100-66-3
ChEBICHEBI:16579
Chemical Name or MaterialAnisole
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only