9-Anthracenecarboxaldehyde, 99%
9-Anthracenecarboxaldehyde, 99%
9-Anthracenecarboxaldehyde, 99%
Thermo Scientific Chemicals

9-Anthracenecarboxaldehyde, 99%

CAS: 642-31-9 | C15H10O | 206.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 153930250
Price (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS642-31-9
IUPAC Nameanthracene-9-carbaldehyde
Molecular FormulaC15H10O
InChI KeyYMNKUHIVVMFOFO-UHFFFAOYSA-N
SMILESO=CC1=C2C=CC=CC2=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or needles
HPLC>=98.5 %
Infrared spectrumConforms
Appearance (Color)Yellow to yellow-green to yellow-brown
Melting point103°C to 107°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only