alpha,alpha'-Dichloro-o-xylene, 98%
alpha,alpha'-Dichloro-o-xylene, 98%
alpha,alpha'-Dichloro-o-xylene, 98%
Thermo Scientific Chemicals

alpha,alpha'-Dichloro-o-xylene, 98%

CAS: 612-12-4 | C8H8Cl2 | 175.05 g/mol
Have Questions?
Catalog NumberQuantity
154141000100 g
Catalog number 154141000
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS612-12-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Semitransparent white to light beige
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
GC>=97.5 %
Melting point53°C to 57°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only