Bromoethane, 98%, pure
Bromoethane, 98%, pure
Bromoethane, 98%, pure
Thermo Scientific Chemicals

Bromoethane, 98%, pure

CAS: 74-96-4 | C2H5Br | 108.966 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 154215000
價格 (TWD)
-
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS74-96-4
IUPAC Namebromoethane
Molecular FormulaC2H5Br
InChI KeyRDHPKYGYEGBMSE-UHFFFAOYSA-N
SMILESCCBr
檢視更多
規格Specification Sheet規格表
GC>=97.5 %
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4225 to 1.4265 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only