3-Bromoquinoline, 98%
3-Bromoquinoline, 98%
3-Bromoquinoline, 98%
Thermo Scientific Chemicals

3-Bromoquinoline, 98%

CAS: 5332-24-1 | C9H6BrN | 208.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 154270250
Price (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5332-24-1
IUPAC Name3-bromoquinoline
Molecular FormulaC9H6BrN
InChI KeyZGIKWINFUGEQEO-UHFFFAOYSA-N
SMILESBrC1=CN=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.6630 to 1.6660 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow to orange
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only