2-Bromothiophene, 98%
2-Bromothiophene, 98%
2-Bromothiophene, 98%
Thermo Scientific Chemicals

2-Bromothiophene, 98%

CAS: 1003-09-4 | C4H3BrS | 163.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1 L
Packaging:
View all
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 154290500
Price (EUR)
39,65
Online exclusive
43,50
Save 3,85 (9%)
Each
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
39,65
Online exclusive
43,50
Save 3,85 (9%)
Each
Chemical Identifiers
CAS1003-09-4
IUPAC Name2-bromothiophene
Molecular FormulaC4H3BrS
InChI KeyTUCRZHGAIRVWTI-UHFFFAOYSA-N
SMILESBrC1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light brown
Infrared spectrumConforms
Refractive index1.5850 to 1.5870 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only