4-tert-Butylcatechol, 99%
4-tert-Butylcatechol, 99%
4-tert-Butylcatechol, 99%
Thermo Scientific Chemicals

4-tert-Butylcatechol, 99%

CAS: 98-29-3 | C10H14O2 | 166.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 154355000
Price (USD)
119.65
Special offer
Online exclusive
Ends: 28-Dec-2026
141.00
Save 21.35
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
119.65
Special offer
Online exclusive
Ends: 28-Dec-2026
141.00
Save 21.35
Each
Chemical Identifiers
CAS98-29-3
IUPAC Name4-tert-butylbenzene-1,2-diol
Molecular FormulaC10H14O2
InChI KeyXESZUVZBAMCAEJ-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to light orange
Appearance (Form)Flakes
Infrared spectrumConforms
Melting point53°C to 57°C
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only