Pierce™ Nickel Coated Plates, Clear, 96-Well

Catalog number: 15442B

Thermo Scientific™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Pierce Nickel Coated Clear 96-Well Plates provide a simple format to bind His-tagged fusion proteins for ELISA and other plate-based assays involving protein interactions of expressed recombinant proteins. These clear plates can be used with colorimetric detection methods.

Features of Pierce Nickel Coated Plates:

• Ni2+ activated surface enables metal-chelate binding of His-tagged proteins
• Detergents used to lyse cells don't inhibit binding to activated plates as they do with plain polystyrene
• Better binding for sensitive assays compared to other commercially available nickel-activated plates
• Detection limit: 1ng of polyhistidine fusion protein
• Binding capacity: approx. 9 pmol His-tagged protein (27kDa) per well
• Activation level: 200 µL

These Ni(2+) chelate-coated plates are ideal for analyzing polyhistidine-tagged fusion proteins by ELISA-based methods. Proteins that contain a succession of several histidine residues at the amino or carboxyl terminus have a strong binding affinity for metal. Bacterial lysates containing polyhistidine-tagged fusion proteins can be added directly to the plates without the need for blocking.

Related Products
Pierce™ Nickel Coated Plates, Clear, 8-Well Strip
Pierce™ Nickel Coated Plates, White, 96-Well
Pierce™ Nickel Coated Plates, Black, 96-Well
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Binding Property: Affinity, Standard Capacity
Color: Clear
Detection Method: Colorimetric
Format: 96-well plate
Plate Blocking: Bovine Serum Albumin (BSA)
Plate Type: Detection Plate, ELISA, Immunoassay
Product Line: Pierce™
Product Size: Custom
Surface Coating: Divalent Nickel, Metal Chelate

Contents & storage

Upon receipt store plates at 4°C in unopened pouches. Once opened, place unused plates in a resealable bag with desiccant and store at 4°C.

Documents

Manuals & protocols