Glycolic acid, 99%
Glycolic acid, 99%
Glycolic acid, 99%
Acros Organics

Glycolic acid, 99%

CAS: 79-14-1 | C2H4O3 | 76.051 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 154515000
HSN 29159090
Price (INR)
38,708.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
38,708.00
Each
Chemical Identifiers
CAS79-14-1
IUPAC Name2-hydroxyacetic acid
Molecular FormulaC2H4O3
InChI KeyAEMRFAOFKBGASW-UHFFFAOYSA-N
SMILESOCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Adhering crystals
Water=<1 %
Melting point72°C to 80°C
Appearance (Color)White to off-white
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only