DL-Camphoroquinone, 99%
DL-Camphoroquinone, 99%
DL-Camphoroquinone, 99%
Thermo Scientific Chemicals

DL-Camphoroquinone, 99%

CAS: 10373-78-1 | C10H14O2 | 166.22 g/mol
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 154570050
Price (JPY)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10373-78-1
IUPAC Name1,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione
Molecular FormulaC10H14O2
InChI KeyVNQXSTWCDUXYEZ-UHFFFAOYNA-N
SMILESCC1(C)C2CCC1(C)C(=O)C2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point199°C to 204°C
GC>=98.5 %
Appearance (Color)Yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only