DL-Camphoroquinone, 99%
DL-Camphoroquinone, 99%
DL-Camphoroquinone, 99%
Thermo Scientific Chemicals

DL-Camphoroquinone, 99%

CAS: 10373-78-1 | C10H14O2 | 166.22 g/mol
数量:
5 g
10 g
50 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 154570500
価格(JPY)
-
数量:
50 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10373-78-1
IUPAC Name1,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione
Molecular FormulaC10H14O2
InChI KeyVNQXSTWCDUXYEZ-UHFFFAOYNA-N
SMILESCC1(C)C2CCC1(C)C(=O)C2=O
さらに表示
Appearance (Form)Crystalline powder
Melting point199°C to 204°C
GC>=98.5 %
Appearance (Color)Yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only