4-Carboxybenzaldehyde, 96%
4-Carboxybenzaldehyde, 96%
4-Carboxybenzaldehyde, 96%
Thermo Scientific Chemicals

4-Carboxybenzaldehyde, 96%

CAS: 619-66-9 | C8H6O3 | 150.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 154581000
Price (EUR)
228,00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
228,00
Each
Chemical Identifiers
CAS619-66-9
IUPAC Name4-formylbenzoic acid
Molecular FormulaC8H6O3
InChI KeyGOUHYARYYWKXHS-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Appearance (Form)Crystalline powder
HPLC>=95 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only