1,4-Cyclohexanedione, 98%
1,4-Cyclohexanedione, 98%
1,4-Cyclohexanedione, 98%
Thermo Scientific Chemicals

1,4-Cyclohexanedione, 98%

CAS: 637-88-7 | C6H8O2 | 112.13 g/mol
数量:
10 g
50 g
250 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 154830100
価格(JPY)
-
数量:
10 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS637-88-7
IUPAC Namecyclohexane-1,4-dione
Molecular FormulaC6H8O2
InChI KeyDCZFGQYXRKMVFG-UHFFFAOYSA-N
SMILESO=C1CCC(=O)CC1
さらに表示
Appearance (Form)Powder or flakes
Melting point76°C to 80°C
Infrared spectrumConforms
Appearance (Color)Yellow to beige
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only