cis-Cyclooctene, 95%, stabilized
cis-Cyclooctene, 95%, stabilized
cis-Cyclooctene, 95%, stabilized
Thermo Scientific Chemicals

cis-Cyclooctene, 95%, stabilized

CAS: 931-87-3 | C8H14 | 110.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 154862500
Price (USD)
78.90
Each
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
78.90
Each
Chemical Identifiers
CAS931-87-3
IUPAC Name(Z)-cyclooctene
Molecular FormulaC8H14
InChI KeyURYYVOIYTNXXBN-UPHRSURJSA-N
SMILESC1CCCC=C/CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4680 to 1.4710 (20°C, 589 nm)
Appearance (Color)Clear colorless to light brown
GC>=94 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only