(+)-Dehydroisoandrosterone, 99%, Thermo Scientific Chemicals
(+)-Dehydroisoandrosterone, 99%, Thermo Scientific Chemicals
(+)-Dehydroisoandrosterone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(+)-Dehydroisoandrosterone, 99%, Thermo Scientific Chemicals

(+)-Dehydroisoandrosterone, CAS # 53-43-0, also known as androstenolone, is an endogenous steroid hormone precursor. It is an intermediate in the conversion of cholesterol to androgens and estrogens.
Have Questions?
變更視圖buttonViewtableView
產品號碼QuantityPackaging
15498050050gGlass bottle
1549800252.5gGlass bottle
15498010010gGlass bottle
產品號碼 154980500
價格 (TWD)
-
Quantity:
50g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS53-43-0
IUPAC Name(3aS,3bR,7S,9aR,9bS,11aS)-7-hydroxy-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-one
Molecular FormulaC19H28O2
InChI KeyFMGSKLZLMKYGDP-USOAJAOKSA-N
SMILESC[C@]12CC[C@H]3[C@@H](CC=C4C[C@@H](O)CC[C@]34C)[C@@H]1CCC2=O
檢視更多
規格Specification Sheet規格表
Appearance (Color)White to off-white
Infrared spectrumConforms
HPLC>=98.5 %
Specific optical rotation+11.0° to +13.0° (20°C, 589 nm) (c=2, Ethanol 96%)
Appearance (Form)Crystalline powder
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• (+)-Dehydroisoandrosterone is an endogenous steroid hormone precursor that acts as an intermediate in the conversion of cholesterol to androgens and estrogens
Applications
• It is an endogenous and naturally occurring steroid hormone produced largely by the adrenal glands, but also by the testes and ovaries
• It has been described that (+)-Dehydroisoandrosterone could have a role in the immune and stress response
• (+)-Dehydroisoandrosterone can be converted to testosterone, androstenedione, estradiol, and estrone
• (+)-Dehydroisoandrosterone is derived from cholesterol through the action of two cytochrome P450 enzymes
• It can induce both direct and indirect in vivo biological effects, by modulating GABA- α receptor and N-Methyl-D-aspartic acid (NMDA) receptor complexes.
• It can also increase pancreatic β cell insulin secretion and anti-glucocorticoid activity
• It is useful to detect excess adrenal activity since it is produced nearly exclusively by the adrenal glands
RUO – Research Use Only
Literature References
Prough, R.; Clark, B.; Klinge, C. Novel mechanisms for DHEA action. Journal of Molecular Endocrinology, 2016, 56, 3, R139-R155.