trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic Acid Monohydrate, 98%
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic Acid Monohydrate, 98%
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic Acid Monohydrate, 98%
Thermo Scientific Chemicals

trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic Acid Monohydrate, 98%

trans-1,2-Diaminocyclohexane-N,N,N,N-tetraacetic acid monohydrate, 98%, C14H24N2O9, CAS Number-125572-95-4 | CAS: 125572-95-4 | C14H20N2O8 | 344.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Catalog number 155000025
Price (USD)
3,277.65
Online exclusive
3,642.00
Save 364.35
Each
Quantity:
2.5 kg
Request bulk or custom format
Price (USD)
3,277.65
Online exclusive
3,642.00
Save 364.35
Each
Chemical Identifiers
CAS125572-95-4
IUPAC Name2-{[(1R,2R)-2-[bis(carboxylatomethyl)azaniumyl]cyclohexyl](carboxylatomethyl)azaniumyl}acetate
Molecular FormulaC14H20N2O8
InChI KeyFCKYPQBAHLOOJQ-NXEZZACHSA-L
SMILES[O-]C(=O)C[NH+](CC([O-])=O)[C@@H]1CCCC[C@H]1[NH+](CC([O-])=O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Loss on drying4.0 to 6.9 % (1g,150°C,6h)
Appearance (Color)White
Infrared spectrumConforms
Sulfated ash=<0.3 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only