Thermo Scientific Chemicals

trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic Acid Monohydrate, 98%, Thermo Scientific Chemicals

Catalog number: 155005000
500 g, Each
Thermo Scientific Chemicals

trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic Acid Monohydrate, 98%, Thermo Scientific Chemicals

Catalog number: 155005000
500 g, Each
Quantity
Catalog number: 155005000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
125572-95-4
IUPAC Name
2-{[(1R,2R)-2-[bis(carboxylatomethyl)azaniumyl]cyclohexyl](carboxylatomethyl)azaniumyl}acetate
Molecular Formula
C14H20N2O8
InChI Key
FCKYPQBAHLOOJQ-NXEZZACHSA-L
SMILES
[O-]C(=O)C[NH+](CC([O-])=O)[C@@H]1CCCC[C@H]1[NH+](CC([O-])=O)CC([O-])=O
Appearance (Color)
White
Infrared spectrum
Conforms
Sulfated ash
=<0.3 %
Appearance (Form)
Crystalline powder
Loss on drying
4.0 to 6.9 % (1g,150°C,6h)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text