UltraPure™ Guanidine Hydrochloride
Invitrogen™

UltraPure™ Guanidine Hydrochloride

UltraPure™ Guanidine Hydrochloride is a strong protein denaturant. In a highly concentrated (5 to 6 M) guanidine hydrochloride solution, manyRead more
Have Questions?
Catalog NumberQuantity
15502016500 g
Catalog number 15502016
Price (USD)
283.65
Online exclusive
295.00
Save 11.35
Each
Quantity:
500 g
Price (USD)
283.65
Online exclusive
295.00
Save 11.35
Each
UltraPure™ Guanidine Hydrochloride is a strong protein denaturant. In a highly concentrated (5 to 6 M) guanidine hydrochloride solution, many proteins unfold and separate into their constituent polypeptide chains. Guanidine hydrochloride is widely used to purify nucleic acids from cell extracts. It is useful as an RNase inhibitor during the isolation of mRNA from mammalian cells.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)RNA Extraction
Packaging TypeBottle
Product LineUltraPure
Product TypeGuanidine Hydrochloride
PurityDNase-Free, RNase-Free
Quantity500 g
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
FormPowder
Unit SizeEach
Contents & Storage
Store at room temperature.