Invitrogen™

UltraPure™ Urea

Catalog number:  15505027

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

UltraPure™ Urea is an effective protein denaturant. In a highly concentrated (6 to 8 M) urea solution, many proteins unfold and separate into their constituent polypeptide chains. UltraPure™ Urea is also used to denature nucleic acids in sequencing gels.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Powder
Product Line: UltraPure™
Product Size: 2 kg
Reagent Type: Urea
Shipping Condition: Approved for shipment at Room Temperature or on Wet Ice

Contents & storage

Storage conditions: 15 – 30°C
Shipping conditions: Ambient or ice
Shelf life: 36 months from date of manufacture

Documents

Manuals & protocols