Search
| 카탈로그 번호 | 수량 |
|---|---|
| 15514011 | 500 mL |
| 15514029 | 3.8 L |
UltraPure Glycerol (≥99% pure) is suitable for concentrating and storing enzymes at low temperatures. It can be diluted to a 50% (v/v) solution that will not freeze at -20°C. With a density of 1.26 g/mL, it is commonly used in electrophoresis loading buffers. Additionally, UltraPure Glycerol gradients are ideal for purifying proteins, bacteriophage, and organelles.
Performance and quality testing: no DNase, RNase, or protease activity detected
For Research Use Only. Not for use in diagnostic procedures.