Product Image
Invitrogen™

UltraPure™ Glycerol

UltraPure Glycerol is suitable for the concentration and storage of enzymes at low temperatures. A 50% (v/v) solution will not freeze at -20°C.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
155140293.8 L
15514011500 mL
Catalog number 15514029
Price (USD)
507.65
Online exclusive
540.00
Save 32.35
Each
Quantity:
3.8 L
Price (USD)
507.65
Online exclusive
540.00
Save 32.35
Each

UltraPure Glycerol (≥99% pure) is suitable for concentrating and storing enzymes at low temperatures. It can be diluted to a 50% (v/v) solution that will not freeze at -20°C. With a density of 1.26 g/mL, it is commonly used in electrophoresis loading buffers. Additionally, UltraPure Glycerol gradients are ideal for purifying proteins, bacteriophage, and organelles.

Performance and quality testing: no DNase, RNase, or protease activity detected

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Chemical Name or MaterialGlycerol
Product LineUltraPure
PurityMolecular Biology Grade
Quantity3.8 L
Shipping ConditionRoom Temperature
FormLiquid
Unit SizeEach
Contents & Storage
Store at room temperature.

Frequently asked questions (FAQs)

What is the storage media for the competent cells?

Preparation procedures and formulations for all of our competent cells are proprietary. All chemically competent cells are delivered in an aqueous solution that contains a mixture of salts, along with a freezing stabilizer such as glycerol or DMSO.