1,2-Difluorobenzene, 98%
1,2-Difluorobenzene, 98%
1,2-Difluorobenzene, 98%
Acros Organics

1,2-Difluorobenzene, 98%

CAS: 367-11-3 | C6H4F2 | 114.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
10 mL
Packaging:
Glass bottle
Catalog number 155150500
also known as 15515
HSN 29039990
Price (INR)
24,238.00
Each
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
24,238.00
Each
Chemical Identifiers
CAS367-11-3
IUPAC Name1,2-difluorobenzene
Molecular FormulaC6H4F2
InChI KeyGOYDNIKZWGIXJT-UHFFFAOYSA-N
SMILESFC1=CC=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)Clear colorless
Refractive index1.4417 to 1.4437 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only