2,3-Dihydroxybenzoic acid, 99%
2,3-Dihydroxybenzoic acid, 99%
2,3-Dihydroxybenzoic acid, 99%
Thermo Scientific Chemicals

2,3-Dihydroxybenzoic acid, 99%

CAS: 303-38-8 | C7H6O4 | 154.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 155170050
Price (USD)
53.65
Online exclusive
59.00
Save 5.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
53.65
Online exclusive
59.00
Save 5.35
Each
Chemical Identifiers
CAS303-38-8
IUPAC Name2,3-dihydroxybenzoic acid
Molecular FormulaC7H6O4
InChI KeyGLDQAMYCGOIJDV-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC(O)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige to pink to grey to brown
Melting point204°C to 208°C
HPLC>=98.5 %
Infrared spectrumConforms
Titration with NaOH>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only