1,4-Diisopropylbenzene, 99%
1,4-Diisopropylbenzene, 99%
1,4-Diisopropylbenzene, 99%
Thermo Scientific Chemicals

1,4-Diisopropylbenzene, 99%

CAS: 100-18-5 | C12H18 | 162.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 155210250
Price (EUR)
37,20
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
37,20
Each
Chemical Identifiers
CAS100-18-5
IUPAC Name1,4-bis(propan-2-yl)benzene
Molecular FormulaC12H18
InChI KeySPPWGCYEYAMHDT-UHFFFAOYSA-N
SMILESCC(C)C1=CC=C(C=C1)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive index1.4875 to 1.4905 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only