3,5-Dimethoxybenzaldehyde, 98%
3,5-Dimethoxybenzaldehyde, 98%
3,5-Dimethoxybenzaldehyde, 98%
Thermo Scientific Chemicals

3,5-Dimethoxybenzaldehyde, 98%

CAS: 7311-34-4 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 155220500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7311-34-4
IUPAC Name3,5-dimethoxybenzaldehyde
Molecular FormulaC9H10O3
InChI KeyVFZRZRDOXPRTSC-UHFFFAOYSA-N
SMILESCOC1=CC(C=O)=CC(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline solid
GC>=97.5 %
Melting point44°C to 48°C
Appearance (Color)White to beige
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only