2,3-Dimethoxyphenol, 99%
2,3-Dimethoxyphenol, 99%
2,3-Dimethoxyphenol, 99%
Thermo Scientific Chemicals

2,3-Dimethoxyphenol, 99%

CAS: 5150-42-5 | C8H10O3 | 154.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 155250250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5150-42-5
IUPAC Name2,3-dimethoxyphenol
Molecular FormulaC8H10O3
InChI KeyQSZCGGBDNYTQHH-UHFFFAOYSA-N
SMILESCOC1=CC=CC(O)=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Appearance (Color)Clear orange to brown
Infrared spectrumConforms
Refractive index1.5385 to 1.5405 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only