(3,4-Dimethoxyphenyl)acetonitrile, 99+%
(3,4-Dimethoxyphenyl)acetonitrile, 99+%
(3,4-Dimethoxyphenyl)acetonitrile, 99+%
Acros Organics

(3,4-Dimethoxyphenyl)acetonitrile, 99+%

CAS: 93-17-4 | C10H11NO2 | 177.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 155260250
also known as 15526
HSN 29269000
Price (INR)
4,793.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
4,793.00
Each
Chemical Identifiers
CAS93-17-4
IUPAC Name2-(3,4-dimethoxyphenyl)acetonitrile
Molecular FormulaC10H11NO2
InChI KeyASLSUMISAQDOOB-UHFFFAOYSA-N
SMILESCOC1=CC=C(CC#N)C=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=99.0 %
Appearance (Color)White to off-white
Appearance (Form)Crystals or crystalline needles
Melting point62°C to 65°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only