UltraPure™ TAE Buffer, 10X
Invitrogen™

UltraPure™ TAE Buffer, 10X

UltraPure™ 10X TAE Buffer is a sterile-filtered solution of 400 mM Tris-acetate and 10 mM EDTA. TAE Buffer is the深入閱讀
Have Questions?
變更視圖buttonViewtableView
產品號碼Quantity
155580421 L
155580264 L
產品號碼 15558042
價格 (TWD)
1,580.00
線上優惠
Ends: 31-Dec-2026
2,250.00
您節省 670.00 (30%)
Each
Quantity:
1 L
Request bulk or custom format
價格 (TWD)
1,580.00
線上優惠
Ends: 31-Dec-2026
2,250.00
您節省 670.00 (30%)
Each

UltraPure™ 10X TAE Buffer is a sterile-filtered solution of 400 mM Tris-acetate and 10 mM EDTA. TAE Buffer is the most commonly used buffer for agarose DNA electrophoresis. It is supplied in 1 L plastic bottles or in a 4 L or 10 L stackable Cubitainer™ Box. A 1X TAE Buffer solution contains 40 mM Tris-acetate and 1 mM EDTA at pH 8.3.

Performance and Quality Testing: No DNase activity detected.

For Research Use Only. Not for use in diagnostic procedures.
規格
BufferRunning Buffer
Concentration10 X
For Use With (Application)Nucleic Acid Gel Electrophoresis, Blotting
Gel CompatibilityAgarose Gels
Product LineUltraPure
Product TypeTAE Buffer
Quantity1 L
Shipping ConditionRoom Temperature
FormatBottle
Unit SizeEach
內容物與存放
Store at room temperature.

常見問答集 (常見問題)

How can generation of replication competent adenoviruses be avoided when using your pSilencer adeno 1.0-CMV System?

Although there is only a very small chance of creating replication competent virus, steps should still be taken to avoid added risk. The virus is expanded in two rounds of amplification, and all secondary expansions should be performed from an initial expansion stock. Secondary amplifications in HEK293 cells should not be performed from the stock of another secondary expansion, as this could increase the chance of making replication competent virus. See the product manual for more detail and guidelines for screening.

Find additional tips, troubleshooting help, and resources within our RNAi Support Center.