N,N,N',N'-Tetraethylethylenediamine, 99+%
N,N,N',N'-Tetraethylethylenediamine, 99+%
N,N,N',N'-Tetraethylethylenediamine, 99+%
Thermo Scientific Chemicals

N,N,N',N'-Tetraethylethylenediamine, 99+%

CAS: 150-77-6 | C10H26N2 | 174.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
100 mL
500 mL
Catalog number 155595000
Price (USD)
-
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS150-77-6
IUPAC Name[2-(diethylazaniumyl)ethyl]diethylazanium
Molecular FormulaC10H26N2
InChI KeyDIHKMUNUGQVFES-UHFFFAOYSA-P
SMILESCC[NH+](CC)CC[NH+](CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=99.0 %
Refractive index1.4333 to 1.4353 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only