Bovine Serum Albumin is suitable for use as an enzyme stabilizer during purification or for dilution of restriction endonucleases andRead more
Have Questions?
Catalog Number
Quantity
15561020
150 mg
Catalog number 15561020
Price (USD)
588,06
Each
Quantity:
150 mg
Request bulk or custom format
Price (USD)
588,06
Each
Bovine Serum Albumin is suitable for use as an enzyme stabilizer during purification or for dilution of restriction endonucleases and nucleic acid modifying enzymes. BSA is also commonly used in DNA and protein labeling experiments as a blocking agent to minimize background. This BSA is acetylated to inactivate contaminating nucleases and proteases. Performance and quality testing Purity is determined by SDS-PAGE. No detectable contaminating activity is observed in 3´ and 5´ exonuclease, DNA nicking, and restriction endonuclease inhibition assays.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeBovine Serum Albumin
ProteinBSA
Quantity150 mg
Shipping ConditionApproved for shipment on Wet or Dry Ice
Concentration45 to 55 mg/mL
Unit SizeEach
Contents & Storage
Bovine Serum Albumin is supplied as an aqueous solution at a concentration of 45 to 55 mg/ml. Store at -20°C. Bovine Serum Albumin is guaranteed for 6 months from date of purchase unless otherwise specified in product literature.
Frequently asked questions (FAQs)
What is the Bovine Serum Albumin (BSA) (Cat. No. 15561020) dissolved in?
BSA is supplied sterile in deionized water.
Citations & References (1)
Citations & References
Abstract
Food deprivation-induced expression of minoxidil sulfotransferase in the hypothalamus uncovered by microarray analysis.
Authors: Li Ji-Yao; Lescure Pascal A; Misek David E; Lai Yu-Mei; Chai Biao-Xin; Kuick Rork; Thompson Robert C; Demo Rebecca M; Kurnit David M; Michailidis George; Hanash Samir M; Gantz Ira;
Journal:J Biol Chem
PubMed ID:11782476
We used oligonucleotide microarrays to analyze comprehensively hypothalamic gene expression changes that correlate with energy homeostasis. We compared the hypothalamic gene expression profiles of freely fed and 48-h fasted rats using 26,379 oligonucleotide probe sets. Expression of 96 genes was up-regulated and expression of 73 genes was down-regulated in a ... More